HCS CellMask™ Deep Red Stain, 1 Set - FAQs
HCS CellMask™ Deep Red Stain, 1 Set - FAQs
View additional product information for HCS CellMask™ Stains - FAQs (H32714, H32713, H32712, H32722, H32721, H32720)
1 product FAQs found
What dyes are used to make the CellMask stains?
The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.