What is the shelf life of the HCS CellMask Near-IR Stain?
As shipped, the product has a warranty period of 1 year from the date of shipment. Once the solid stain is resolubilized in DMSO, it should be stored frozen, desiccated, and protected from light and is useable for ~ 6 months.