2-Fluoro-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals
2-Fluoro-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals
2-Fluoro-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals
2-Fluoro-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H32724.06
also known as H32724-06
5 g
Catalog number H32724.06
also known as H32724-06
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.90
Save 11.25 (14%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.90
Save 11.25 (14%)
Each
Add to cart
Chemical Identifiers
CAS127667-01-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Melting Point (clear melt)78.5-84.5?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only