2-Fluoro-6-methoxybenzamide, 98%
2-Fluoro-6-methoxybenzamide, 98%
2-Fluoro-6-methoxybenzamide, 98%
Thermo Scientific Chemicals

2-Fluoro-6-methoxybenzamide, 98%

CAS: 529512-81-0 | C8H8FNO2 | 169.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H32774.03
also known as H32774-03
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Chemical Identifiers
CAS529512-81-0
IUPAC Name2-fluoro-6-methoxybenzamide
Molecular FormulaC8H8FNO2
InChI KeyBDJNFURQDZEPLK-UHFFFAOYSA-N
SMILESCOC1=C(C(N)=O)C(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (GC)≥97.5%
Melting Point (clear melt)190.0-196.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only