(+/-)-1-Boc-pyrrolidine-2-methanol, 98%
(+/-)-1-Boc-pyrrolidine-2-methanol, 98%
(+/-)-1-Boc-pyrrolidine-2-methanol, 98%
Thermo Scientific Chemicals

(+/-)-1-Boc-pyrrolidine-2-methanol, 98%

CAS: 170491-63-1 | C10H19NO3 | 201.266 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32786.03
also known as H32786-03
Price (USD)
34.65
Online Exclusive
38.40
Save 3.75 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
38.40
Save 3.75 (10%)
Each
Chemical Identifiers
CAS170491-63-1
IUPAC Nametert-butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular FormulaC10H19NO3
InChI KeyBFFLLBPMZCIGRM-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (GC)≥97.5%
Melting Point (clear melt)50.5-56.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only