2-Amino-5-phenyl-1,3,4-thiadiazole, 97%
Thermo Scientific Chemicals

2-Amino-5-phenyl-1,3,4-thiadiazole, 97%

CAS: 2002-03-1 | C7H6N4S | 178.21 g/mol
Have Questions?
Catalog NumberQuantity
H32826.03
also known as H32826-03
1 g
Catalog number H32826.03
also known as H32826-03
Price (USD)
34.65
Online Exclusive
37.90
Save 3.25 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
37.90
Save 3.25 (9%)
Each
Chemical Identifiers
CAS2002-03-1
IUPAC Name5-phenyl-1,3,4-thiadiazol-2-amine
Molecular FormulaC8H7N3S
InChI KeyUHZHEOAEJRHUBW-UHFFFAOYSA-N
SMILESNC1=NN=C(S1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥96.0%
Melting Point (clear melt)227.5-233.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only