Boron trichloride, 1M soln. in hexanes, mixed isomers
Boron trichloride, 1M soln. in hexanes, mixed isomers
Boron trichloride, 1M soln. in hexanes, mixed isomers
Thermo Scientific Chemicals

Boron trichloride, 1M soln. in hexanes, mixed isomers

CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number H32833.AP
also known as H32833-AP
Price (USD)
225.65
Special offer
Online exclusive
Ends: 30-Jun-2026
265.00
Save 39.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
225.65
Special offer
Online exclusive
Ends: 30-Jun-2026
265.00
Save 39.35 (15%)
Each
Chemical Identifiers
CAS10294-34-5
IUPAC Nametrichloroborane
Molecular FormulaBCl3
InChI KeyFAQYAMRNWDIXMY-UHFFFAOYSA-N
SMILESClB(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Appearance (Color)Colourless
Assay (Titration ex Chloride)0.9 - 1.1M
A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat. Unstable. Store at 2-8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. F.M. Dean,; J.Goodchild,; L.E. Houghton,; J.A. Martin,; R.B. Morton,; B. Parton,; A.W. Price, ;Nongyow Somvichien. Boron trichloride as a selective demethylating agent. Tetrahedron Letters. 1996, 7(35),4153-4159.
  2. N. N. Greenwood,; K. Wade .Complexes of boron trichloride with pyridine and piperidine. J. Chem. Soc., year of publication , 1130-1141.