Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H32847.03
also known as H32847-03
1 g
Catalog number H32847.03
also known as H32847-03
Price (USD)
142.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
142.00
Each
Chemical Identifiers
CAS1040683-08-6
IUPAC Nameethyl 1-(trifluoromethyl)cyclobutane-1-carboxylate
Molecular FormulaC8H11F3O2
InChI KeyAYUISFJYNKIHDD-UHFFFAOYSA-N
SMILESCCOC(=O)C1(CCC1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3795-1.3845 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only