3-Phenoxybenzeneboronic acid, 97+%
3-Phenoxybenzeneboronic acid, 97+%
3-Phenoxybenzeneboronic acid, 97+%
Thermo Scientific Chemicals

3-Phenoxybenzeneboronic acid, 97+%

CAS: 221006-66-2 | C12H11BO3 | 214.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32879.03
also known as H32879-03
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Chemical Identifiers
CAS221006-66-2
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)135.0-144.0?C
Appearance (Color)White
Assay (HPLC)≥97.0%
Proton NMRConforms to structure
FormPowder
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S Schmidt,; R M Wittich,; D Erdmann,; H Wilkes,; W Francke,; P Fortnagel. Biodegradation of diphenyl ether and its monohalogenated derivatives by Sphingomonas sp. strain SS3.. Appl. Environ. Microbiol. 1982, 58 (9),2744-2750.
  2. U Örn, L Eriksson,; E Jakobsson. Synthesis and characterization of polybrominated diphenyl ethers-unlabelled and radiolabelled tetra-, penta-and hexa-bromodiphenyl ethers . Acta Chemica Scandinavia. 1996, 50 802-807.