4-Acetylbenzenesulfonamide, 97%
4-Acetylbenzenesulfonamide, 97%
4-Acetylbenzenesulfonamide, 97%
Thermo Scientific Chemicals

4-Acetylbenzenesulfonamide, 97%

CAS: 1565-17-9 | C8H9NO3S | 199.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32901.03
also known as H32901-03
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS1565-17-9
IUPAC Name4-acetylbenzene-1-sulfonamide
Molecular FormulaC8H9NO3S
InChI KeyCSATVXJBGFVJES-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White to yellow
Assay (HPLC)≥96.0%
Melting Point (clear melt)174.0-183.0?C
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

Solubility
soluble in water( 2650 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Sachiko Nagamine,; Tomoko Otawa,; Hiroko Nakai,; Shozo Asada. Estimation of the Rates of Available Fraction for Some 4-Substituted Acetophenone Derivatives in the Rat Reversible Drug-Metabolite Pharmacokinetics. Chemical and Pharmaceutical Bulletin. 1988, 36(11), 4612-4618.
  2. Aziz-ur-Rehmana,; Shahid Rasoola,; Muhammad Athar Abbasia,; Ambreen Fatimaa,; Khadija Nafeesaa,; Irshad Ahmadb,; Saira Afzalb.Synthesis, spectral analysis and biological screening of some new N-(un)substituted N-(5-chloro-2-methoxyphenyl)-aryl sulfonamides. Journal of Pharmacy Research. 2013, 6(5) ,559-564.