2-Chloro-4-fluoro-1-nitrobenzene, 98%
2-Chloro-4-fluoro-1-nitrobenzene, 98%
2-Chloro-4-fluoro-1-nitrobenzene, 98%
Thermo Scientific Chemicals

2-Chloro-4-fluoro-1-nitrobenzene, 98%

CAS: 2106-50-5 | C6H3ClFNO2 | 175.543 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H32906.06
also known as H32906-06
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS2106-50-5
IUPAC Name2-chloro-4-fluoro-1-nitrobenzene
Molecular FormulaC6H3ClFNO2
InChI KeyKQOOFMWRLDRDAX-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to yellow-brown
FormFused solid or lumps or crystals or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)32-37?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only