4-(Hydroxymethyl)pyridine-2-carboxylic acid, 95%
4-(Hydroxymethyl)pyridine-2-carboxylic acid, 95%
4-(Hydroxymethyl)pyridine-2-carboxylic acid, 95%
Thermo Scientific Chemicals

4-(Hydroxymethyl)pyridine-2-carboxylic acid, 95%

CAS: 923169-37-3 | C7H7NO3 | 153.137 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H32921.03
also known as H32921-03
Price (USD)
52.65
Online Exclusive
58.30
Save 5.65 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
52.65
Online Exclusive
58.30
Save 5.65 (10%)
Each
Chemical Identifiers
CAS923169-37-3
IUPAC Name4-(hydroxymethyl)pyridine-2-carboxylic acid
Molecular FormulaC7H7NO3
InChI KeyQMLYYPGJXCVEHA-UHFFFAOYSA-N
SMILESOCC1=CC(=NC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown or pale pink
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
FormPowder
Assay (HPLC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only