1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester, 95%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester, 95%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester, 95%

CAS: 844891-04-9 | C12H21BN2O2 | 236.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32930.03
also known as H32930-03
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS844891-04-9
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only