1-Bromo-4-n-dodecylbenzene, 98%
1-Bromo-4-n-dodecylbenzene, 98%
1-Bromo-4-n-dodecylbenzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-n-dodecylbenzene, 98%

CAS: 126930-72-1 | C18H29Br | 325.334 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32963.14
also known as H32963-14
Price (USD)
646.65
Online Exclusive
718.00
Save 71.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
646.65
Online Exclusive
718.00
Save 71.35 (10%)
Each
Chemical Identifiers
CAS126930-72-1
IUPAC Name1-bromo-4-dodecylbenzene
Molecular FormulaC18H29Br
InChI KeyRFFMRBISTZHCNY-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5035-1.5075 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only