3,3'-Diethyloxadicarbocyanine iodide, 96%
3,3'-Diethyloxadicarbocyanine iodide, 96%
3,3'-Diethyloxadicarbocyanine iodide, 96%
Thermo Scientific Chemicals

3,3'-Diethyloxadicarbocyanine iodide, 96%

CAS: 14806-50-9 | C23H23IN2O2 | 486.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H33094.03
also known as H33094-03
Price (USD)
118.65
Online exclusive
132.00
Save 13.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
118.65
Online exclusive
132.00
Save 13.35
Each
Chemical Identifiers
CAS14806-50-9
SpecificationsSpecification SheetSpecification Sheet
Assay (special)≥95.0 to ≤105.0% (based on elemental I and N)
Elemental AnalysisN: 5.47-6.05% (Theory: 5.76%)
Appearance (Color)Dark blue
Elemental AnalysisI: 24.79-27.40% (Theory: 26.09%)
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only