4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta-4,5-thieno[2,3-d]pyrimidine, 96%
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta-4,5-thieno[2,3-d]pyrimidine, 96%
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta-4,5-thieno[2,3-d]pyrimidine, 96%
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta-4,5-thieno[2,3-d]pyrimidine, 96%
Thermo Scientific Chemicals

4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta-4,5-thieno[2,3-d]pyrimidine, 96%

CAS: 40106-58-9 | C11H11ClN2S | 238.733 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33184.03
also known as H33184-03
Price (USD)
60.65
Online Exclusive
66.80
Save 6.15 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
66.80
Save 6.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS40106-58-9
IUPAC Name3-chloro-8-thia-4,6-diazatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraene
Molecular FormulaC11H11ClN2S
InChI KeyAUYMUMFPLSANID-UHFFFAOYSA-N
SMILESClC1=C2C3=C(CCCCC3)SC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (GC)≥95.0%
Melting Point (clear melt)60.0-69.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only