2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%
2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%
2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%
2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%
Thermo Scientific Chemicals

2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%

CAS: 1227628-83-2 | C8H4F4O2 | 208.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33203.03
also known as H33203-03
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1227628-83-2
IUPAC Name2-fluoro-4-(trifluoromethoxy)benzaldehyde
Molecular FormulaC8H4F4O2
InChI KeyDEACWPDDLIOZJV-UHFFFAOYSA-N
SMILESFC1=C(C=O)C=CC(OC(F)(F)F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Free acid (titration)≤1.5%
Refractive Index1.4430-1.4480 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only