3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
Thermo Scientific Chemicals

3-Acetylquinoline, 97%

CAS: 33021-53-3 | C11H9NO | 171.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33206.03
also known as H33206-03
Price (USD)
83.65
Special offer
Online exclusive
Ends: 30-Jun-2026
98.10
Save 14.45 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 30-Jun-2026
98.10
Save 14.45 (15%)
Each
Add to cart
Chemical Identifiers
CAS33021-53-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormPowder
Melting Point (clear melt)94.0-104.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only