3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
3-Acetylquinoline, 97%
Thermo Scientific Chemicals

3-Acetylquinoline, 97%

CAS: 33021-53-3 | C11H9NO | 171.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33206.06
also known as H33206-06
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS33021-53-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormPowder
Melting Point (clear melt)94.0-104.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only