2-(Trifluoromethyl)pyrazine, 97%
2-(Trifluoromethyl)pyrazine, 97%
2-(Trifluoromethyl)pyrazine, 97%
2-(Trifluoromethyl)pyrazine, 97%
Thermo Scientific Chemicals

2-(Trifluoromethyl)pyrazine, 97%

CAS: 61655-67-2 | C5H3F3N2 | 148.088 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33338.03
also known as H33338-03
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS61655-67-2
IUPAC Name2-(trifluoromethyl)pyrazine
Molecular FormulaC5H3F3N2
InChI KeyCEAMSISEJZMQEP-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4145-1.4195 @ 20?C
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only