1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%
Thermo Scientific Chemicals

1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%

CAS: 1215564-15-0 | C8H4ClF3N4O2S | 312.651 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33424.MD
also known as H33424-MD
Price (USD)
31.60
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
31.60
Each
Add to cart
Chemical Identifiers
CAS1215564-15-0
IUPAC Name1-[4-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride
Molecular FormulaC8H4ClF3N4O2S
InChI KeySADUDBJMXGGXDE-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NC(=NC=C1)N1C=C(C=N1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Assay (GC)≥94.0%
Melting Point (clear melt)92.0-105.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only