2-Amino-3,3,3-trifluoropropionic acid, 97%
2-Amino-3,3,3-trifluoropropionic acid, 97%
2-Amino-3,3,3-trifluoropropionic acid, 97%
2-Amino-3,3,3-trifluoropropionic acid, 97%
Thermo Scientific Chemicals

2-Amino-3,3,3-trifluoropropionic acid, 97%

CAS: 17463-43-3 | C3H4F3NO2 | 143.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33461.03
also known as H33461-03
Price (USD)
381.65
Special offer
Online exclusive
Ends: 30-Jun-2026
449.00
Save 67.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
381.65
Special offer
Online exclusive
Ends: 30-Jun-2026
449.00
Save 67.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS17463-43-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only