6-Chloro-4-hydroxy-2-methylpyrimidine, 97%
6-Chloro-4-hydroxy-2-methylpyrimidine, 97%
6-Chloro-4-hydroxy-2-methylpyrimidine, 97%
Thermo Scientific Chemicals

6-Chloro-4-hydroxy-2-methylpyrimidine, 97%

CAS: 17551-52-9 | C5H5ClN2O | 144.558 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H33478.03
also known as H33478-03
Price (USD)
76.65
Online Exclusive
84.50
Save 7.85 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
76.65
Online Exclusive
84.50
Save 7.85 (9%)
Each
Chemical Identifiers
CAS17551-52-9
IUPAC Name6-chloro-2-methyl-1,4-dihydropyrimidin-4-one
Molecular FormulaC5H5ClN2O
InChI KeyQNANRGHPXMUJQC-UHFFFAOYSA-N
SMILESCC1=NC(=O)C=C(Cl)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormPowder
Appearance (Color)White
Melting Point (clear melt)228.0-238.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only