1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%
1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%
1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%
Thermo Scientific Chemicals

1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%

CAS: 36321-73-0 | C10H12Br2 | 292.014 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33558.03
also known as H33558-03
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 30-Jun-2026
119.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS36321-73-0
IUPAC Name1,2-dibromo-3,4,5,6-tetramethylbenzene
Molecular FormulaC10H12Br2
InChI KeyVEZJOZSIGSNYEB-UHFFFAOYSA-N
SMILESCC1=C(C)C(C)=C(Br)C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)206.0-212.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only