3-Cyanopyridazine, 97%
3-Cyanopyridazine, 97%
3-Cyanopyridazine, 97%
Thermo Scientific Chemicals

3-Cyanopyridazine, 97%

CAS: 53896-49-4 | C5H3N3 | 105.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33568.03
also known as H33568-03
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS53896-49-4
IUPAC Namepyridazine-3-carbonitrile
Molecular FormulaC5H3N3
InChI KeyPJESVVYWPFAJCS-UHFFFAOYSA-N
SMILESN#CC1=NN=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Appearance (Color)Cream to brown
Melting Point (clear melt)41.0-51.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only