2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%
2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%
2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%
Thermo Scientific Chemicals

2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%

CAS: 115010-11-2 | C8H7ClO3S | 218.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Catalog number H33579.06
also known as H33579-06
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS115010-11-2
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)79.5-88.5?C
Appearance (Color)White to pale cream
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only