2-Phenoxybenzyl bromide, 97%
2-Phenoxybenzyl bromide, 97%
2-Phenoxybenzyl bromide, 97%
Thermo Scientific Chemicals

2-Phenoxybenzyl bromide, 97%

CAS: 82657-72-5 | C13H11BrO | 263.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H33585.MD
also known as H33585-MD
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS82657-72-5
IUPAC Name1-(bromomethyl)-2-phenoxybenzene
Molecular FormulaC13H11BrO
InChI KeyYQRIQBOWLXRKKG-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear pale yellow
Refractive Index1.6055-1.6105 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only