3-(2-Chloro-6-fluorophenyl)propionic acid, 96%
3-(2-Chloro-6-fluorophenyl)propionic acid, 96%
3-(2-Chloro-6-fluorophenyl)propionic acid, 96%
Thermo Scientific Chemicals

3-(2-Chloro-6-fluorophenyl)propionic acid, 96%

CAS: 88740-77-6 | C9H8ClFO2 | 202.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33586.06
also known as H33586-06
Price (USD)
284.65
Online Exclusive
316.00
Save 31.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
284.65
Online Exclusive
316.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS88740-77-6
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥95.0%
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Melting Point (clear melt)54.5-63.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only