4-Acetyl-2-chloropyridine, 97%
4-Acetyl-2-chloropyridine, 97%
4-Acetyl-2-chloropyridine, 97%
4-Acetyl-2-chloropyridine, 97%
Thermo Scientific Chemicals

4-Acetyl-2-chloropyridine, 97%

CAS: 23794-15-2 | C7H6ClNO | 155.581 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33619.03
also known as H33619-03
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
120.00
Save 17.35 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
120.00
Save 17.35 (14%)
Each
Add to cart
Chemical Identifiers
CAS23794-15-2
IUPAC Name1-(2-chloropyridin-4-yl)ethan-1-one
Molecular FormulaC7H6ClNO
InChI KeyZJCGPQZERFBGSM-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(Cl)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)33.5-42.5?C
Assay (GC)≥96.0%
Appearance (Color)White to pale cream to yellow
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only