Rhodamine 6G perchlorate, 96%
Rhodamine 6G perchlorate, 96%
Rhodamine 6G perchlorate, 96%
Thermo Scientific Chemicals

Rhodamine 6G perchlorate, 96%

CAS: 13161-28-9 | C28H31ClN2O7 | 543.013 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33626.MD
also known as H33626-MD
Price (USD)
38.65
Online Exclusive
42.90
Save 4.25 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.90
Save 4.25 (10%)
Each
Chemical Identifiers
CAS13161-28-9
IUPAC Name9-[2-(ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethyl-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC28H31ClN2O3
InChI KeyXFKVYXCRNATCOO-UHFFFAOYSA-M
SMILES[Cl-].CCNC1=CC2=[O+]C3=CC(NCC)=C(C)C=C3C(C3=CC=CC=C3C(=O)OCC)=C2C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to red to dark red
Elemental AnalysisC: 58.84-65.03% (Theory: 61.94%)
Elemental AnalysisH: 5.46-6.04% (Theory: 5.75%)
Assay (special)≥95.0 to ≤105.0% (based on elemental C, H, Cl)
Elemental AnalysisCl: 6.20-6.86% (Theory: 6.53%)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only