9-Methylacridine, 96%
9-Methylacridine, 96%
9-Methylacridine, 96%
Thermo Scientific Chemicals

9-Methylacridine, 96%

CAS: 611-64-3 | C14H11N | 193.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33718.03
also known as H33718-03
Price (USD)
145.65
Special offer
Online exclusive
Ends: 28-Dec-2026
171.00
Save 25.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
145.65
Special offer
Online exclusive
Ends: 28-Dec-2026
171.00
Save 25.35
Each
Chemical Identifiers
CAS611-64-3
IUPAC Name9-methylacridine
Molecular FormulaC14H11N
InChI KeyFLDRLXJNISEWNZ-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormPowder or crystalline powder
Assay (HPLC)≥95.0%
Identification (FTIR)Conforms
Melting Point (clear melt)111.0-120.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only