3-[4-(Benzyloxy)phenyl]propionic acid, 96%
3-[4-(Benzyloxy)phenyl]propionic acid, 96%
3-[4-(Benzyloxy)phenyl]propionic acid, 96%
3-[4-(Benzyloxy)phenyl]propionic acid, 96%
Thermo Scientific Chemicals

3-[4-(Benzyloxy)phenyl]propionic acid, 96%

CAS: 50463-48-4 | C16H16O3 | 256.301 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33766.06
also known as H33766-06
Price (USD)
93.60
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
93.60
Each
Add to cart
Chemical Identifiers
CAS50463-48-4
IUPAC Name3-[4-(benzyloxy)phenyl]propanoic acid
Molecular FormulaC16H16O3
InChI KeyQTSAUVQZNADEKS-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (Silylated GC)≥95.0%
Melting Point (clear melt)115.0-125.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only