2-Methylbenzothiazol-6-ol, 96%
2-Methylbenzothiazol-6-ol, 96%
2-Methylbenzothiazol-6-ol, 96%
Thermo Scientific Chemicals

2-Methylbenzothiazol-6-ol, 96%

CAS: 68867-18-5 | C8H7NOS | 165.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33812.03
also known as H33812-03
Price (USD)
99.65
Online exclusive
110.00
Save 10.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
99.65
Online exclusive
110.00
Save 10.35
Each
Chemical Identifiers
CAS68867-18-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale gray
Melting Point (clear melt)156.0-165.0?C
Assay (GC)≥95.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only