3-Fluoro-2-nitropyridine, 96%
3-Fluoro-2-nitropyridine, 96%
3-Fluoro-2-nitropyridine, 96%
Thermo Scientific Chemicals

3-Fluoro-2-nitropyridine, 96%

CAS: 54231-35-5 | C5H3FN2O2 | 142.089 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33813.03
also known as H33813-03
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
150.65
Online Exclusive
167.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS54231-35-5
IUPAC Name3-fluoro-2-nitropyridine
Molecular FormulaC5H3FN2O2
InChI KeyIJVFHCSUEBAAOZ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=C(F)C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to dark brown
Assay (GC)≥95.0%
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)28.5-37.5?C
A specialty product for proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A specialty product for proteomics research applications.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Louis A. Carpino. 1-Hydroxy-7-azabenzotriazole. An efficient peptide coupling additive. J. Am. Chem. Soc. 1993115 (10), 4397-4398
  2. Ivano Messina; Igor Popa; Vítězslav Maier; and Miroslav Soural.Solid-Phase Synthesis of 5-Noranagrelide Derivatives. ACS Comb. Sci. 201416 (1), 33-38