Methyl 2-iodo-5-methylbenzoate, 95%
Methyl 2-iodo-5-methylbenzoate, 95%
Methyl 2-iodo-5-methylbenzoate, 95%
Thermo Scientific Chemicals

Methyl 2-iodo-5-methylbenzoate, 95%

CAS: 103440-52-4 | C9H9IO2 | 276.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33853.06
also known as H33853-06
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS103440-52-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.5920-1.5980 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only