2-Chloro-5-(trifluoromethyl)pyrazine, 97%
2-Chloro-5-(trifluoromethyl)pyrazine, 97%
2-Chloro-5-(trifluoromethyl)pyrazine, 97%
Thermo Scientific Chemicals

2-Chloro-5-(trifluoromethyl)pyrazine, 97%

CAS: 799557-87-2 | C5H2ClF3N2 | 182.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33863.MD
also known as H33863-MD
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Chemical Identifiers
CAS799557-87-2
IUPAC Name2-chloro-5-(trifluoromethyl)pyrazine
Molecular FormulaC5H2ClF3N2
InChI KeyAIEGIFIEQXZBCP-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CN=C(Cl)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4440-1.4490 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only