4-Phenoxybenzyl bromide, 97%
4-Phenoxybenzyl bromide, 97%
4-Phenoxybenzyl bromide, 97%
Thermo Scientific Chemicals

4-Phenoxybenzyl bromide, 97%

CAS: 36881-42-2 | C13H11BrO | 263.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33895.MD
also known as H33895-MD
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS36881-42-2
IUPAC Name1-(bromomethyl)-4-phenoxybenzene
Molecular FormulaC13H11BrO
InChI KeyCPIGBCFBFZSCQI-UHFFFAOYSA-N
SMILESBrCC1=CC=C(OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6140-1.6190 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only