2-Hydroxy-4-methylbenzaldehyde, 97+%
2-Hydroxy-4-methylbenzaldehyde, 97+%
2-Hydroxy-4-methylbenzaldehyde, 97+%
Thermo Scientific Chemicals

2-Hydroxy-4-methylbenzaldehyde, 97+%

CAS: 698-27-1 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H33909.06
also known as H33909-06
Price (USD)
88.65
Online Exclusive
98.10
Save 9.45 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.10
Save 9.45 (10%)
Each
Chemical Identifiers
CAS698-27-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to pale brown or pale pink
Assay (GC)≥97.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)55.0-65.0?C
Assay (HPLC)≥97.0%
The intermediate for synthesis of liquid crystal. Also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The intermediate for synthesis of liquid crystal. Also used as pharmaceutical intermediate.

Solubility
Soluble in oil, slightly soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents. Store under inert gas.
RUO – Research Use Only

General References:

  1. N. E. Eltaye; S. G. Teoh; J. B.-J. Teh; H.-K. Fun and K. Ibrahim. 2,2'-[1,2-Phenylenebis(nitrilomethylidyne)]bis(5-methylphenol). Acta Cryst. 2007, E63, o695-o696.
  2. Arthur Clayton, B.Sc., A.R.C.S. CXXVIII.—The action of alkalis on certain derivatives of coumarin. J. Chem. Soc., Trans. 1910, 97, 1388-1408.