7-Bromo-5-fluorobenzo[b]furan, 97%
7-Bromo-5-fluorobenzo[b]furan, 97%
7-Bromo-5-fluorobenzo[b]furan, 97%
Thermo Scientific Chemicals

7-Bromo-5-fluorobenzo[b]furan, 97%

CAS: 253429-19-5 | C8H4BrFO | 215.021 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33948.MD
also known as H33948-MD
Price (USD)
79.65
Online Exclusive
88.60
Save 8.95 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
79.65
Online Exclusive
88.60
Save 8.95 (10%)
Each
Chemical Identifiers
CAS253429-19-5
IUPAC Name7-bromo-5-fluoro-1-benzofuran
Molecular FormulaC8H4BrFO
InChI KeyTYBMMMOEVBNTDU-UHFFFAOYSA-N
SMILESFC1=CC(Br)=C2OC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5780-1.5830 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only