n-Heptane, 95%
n-Heptane, 95%
n-Heptane, 95%
Thermo Scientific Chemicals

n-Heptane, 95%

CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
2500 mL
Catalog number H33987.0F
also known as H33987-0F
Price (USD)
79.65
Online Exclusive
87.80
Save 8.15 (9%)
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
79.65
Online Exclusive
87.80
Save 8.15 (9%)
Each
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.3850-1.3910 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

Solubility
Miscible with alcohol, chloroform, and ether. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only

General References:

  1. Caponetti, E.; Pedone, L.; Martino, D. C.; Panto, V.; Liveri, V. T. Synthesis, size control, and passivation of CdS nanoparticles in water/AOT-heptane microemulsions. Mater. Sci. Eng. C 2003, 23 (4), 531-539.
  2. Chica, A.; Corma, A. Hydroisomerization of Pentane, Hexane, and Heptane for Improving the Octane Number of Gasoline. J. Catal. 1999, 187 (1), 167-176.