2-Methyl-1,8-naphthyridine, 97%, Thermo Scientific Chemicals
2-Methyl-1,8-naphthyridine, 97%, Thermo Scientific Chemicals
2-Methyl-1,8-naphthyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-1,8-naphthyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H34032.03
also known as H34032-03
1 g
Catalog number H34032.03
also known as H34032-03
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS1569-16-0
IUPAC Name2-methyl-1,8-naphthyridine
Molecular FormulaC9H8N2
InChI KeyFSWRUYCICUXURT-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=NC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤2.25%
Melting Point (clear melt)91.0-100.0?C
Appearance (Color)Orange to brown
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only