Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%
Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%
Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%
Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%
Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%
Thermo Scientific Chemicals

Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%

CAS: 761441-54-7 | C6F3LiN4 | 192.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H34081.03
also known as H34081-03
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS761441-54-7
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
Identification (FTIR)Conforms
Carbon-13 NMRConforms to structure
Appearance (Color)White to cream or pale yellow
FormPowder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only