2-Fluoro-4-(pentafluorothio)aniline, 97%
2-Fluoro-4-(pentafluorothio)aniline, 97%
2-Fluoro-4-(pentafluorothio)aniline, 97%
Thermo Scientific Chemicals

2-Fluoro-4-(pentafluorothio)aniline, 97%

CAS: 1240257-25-3 | C6H5F6NS | 237.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34099.MD
also known as H34099-MD
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Chemical Identifiers
CAS1240257-25-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to very pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4690-1.4740 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only