6-(1H-Pyrazol-1-yl)nicotinic acid, 97%
6-(1H-Pyrazol-1-yl)nicotinic acid, 97%
6-(1H-Pyrazol-1-yl)nicotinic acid, 97%
6-(1H-Pyrazol-1-yl)nicotinic acid, 97%
Thermo Scientific Chemicals

6-(1H-Pyrazol-1-yl)nicotinic acid, 97%

CAS: 253315-22-9 | C9H7N3O2 | 189.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34104.03
also known as H34104-03
Price (USD)
62.65
Online Exclusive
69.60
Save 6.95 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
69.60
Save 6.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS253315-22-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)266.5-275.5?C
Assay (Silylated GC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only