3-(2-Fluorophenyl)propionic acid, 96%
3-(2-Fluorophenyl)propionic acid, 96%
3-(2-Fluorophenyl)propionic acid, 96%
Thermo Scientific Chemicals

3-(2-Fluorophenyl)propionic acid, 96%

CAS: 1643-26-1 | C9H9FO2 | 168.167 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H34110.03
also known as H34110-03
Price (USD)
35.65
Online Exclusive
39.40
Save 3.75 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.40
Save 3.75 (10%)
Each
Chemical Identifiers
CAS1643-26-1
IUPAC Name3-(2-fluorophenyl)propanoate
Molecular FormulaC9H8FO2
InChI KeyGUZLQEOSDXLCKX-UHFFFAOYSA-M
SMILES[O-]C(=O)CCC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (Silylated GC)≥95.0%
FormPowder
Melting Point (clear melt)72.5-81.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only