3-(Difluoromethyl)aniline, 97%
3-(Difluoromethyl)aniline, 97%
3-(Difluoromethyl)aniline, 97%
Thermo Scientific Chemicals

3-(Difluoromethyl)aniline, 97%

CAS: 368-99-0 | C7H7F2N | 143.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34188.03
also known as H34188-03
Price (USD)
281.65
Online exclusive
313.00
Save 31.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
281.65
Online exclusive
313.00
Save 31.35
Each
Chemical Identifiers
CAS368-99-0
IUPAC Name3-(difluoromethyl)aniline
Molecular FormulaC7H7F2N
InChI KeyIDFPXKDJNDYDKA-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)C(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5210-1.5260 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only