2-Amino-3-cyano-5-iodopyridine, 97%
2-Amino-3-cyano-5-iodopyridine, 97%
2-Amino-3-cyano-5-iodopyridine, 97%
Thermo Scientific Chemicals

2-Amino-3-cyano-5-iodopyridine, 97%

CAS: 1347815-41-1 | C6H4IN3 | 245.023 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H34191.06
also known as H34191-06
Price (USD)
267.65
Online Exclusive
297.00
Save 29.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
267.65
Online Exclusive
297.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS1347815-41-1
IUPAC Name2-amino-5-iodopyridine-3-carbonitrile
Molecular FormulaC6H4IN3
InChI KeyNJLCSLOABAUGHW-UHFFFAOYSA-N
SMILESNC1=NC=C(I)C=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)192.0-198.0?C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only