2-(4-Fluorophenoxy)benzaldehyde, 97%
2-(4-Fluorophenoxy)benzaldehyde, 97%
2-(4-Fluorophenoxy)benzaldehyde, 97%
Thermo Scientific Chemicals

2-(4-Fluorophenoxy)benzaldehyde, 97%

CAS: 320423-61-8 | C13H9FO2 | 216.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34259.03
also known as H34259-03
Price (USD)
77.65
Online Exclusive
86.60
Save 8.95 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
86.60
Save 8.95 (10%)
Each
Chemical Identifiers
CAS320423-61-8
IUPAC Name2-(4-fluorophenoxy)benzaldehyde
Molecular FormulaC13H9FO2
InChI KeyRZJFZXRBXCJPTA-UHFFFAOYSA-N
SMILESFC1=CC=C(OC2=CC=CC=C2C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (GC)≤96.0%
Melting Point (clear melt)51-56?C
Free acid (titration)≤2.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only