2-Aminooxazole, 97%
2-Aminooxazole, 97%
2-Aminooxazole, 97%
Thermo Scientific Chemicals

2-Aminooxazole, 97%

CAS: 4570-45-0 | C3H4N2O | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H34360.06
also known as H34360-06
Price (USD)
446.65
Online Exclusive
496.00
Save 49.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
446.65
Online Exclusive
496.00
Save 49.35 (10%)
Each
Chemical Identifiers
CAS4570-45-0
IUPAC Name1,3-oxazol-2-amine
Molecular FormulaC3H4N2O
InChI KeyACTKAGSPIFDCMF-UHFFFAOYSA-N
SMILESNC1=NC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)91.0-100.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only