3-(2-Bromophenyl)-1H-pyrazole, 97%
3-(2-Bromophenyl)-1H-pyrazole, 97%
3-(2-Bromophenyl)-1H-pyrazole, 97%
Thermo Scientific Chemicals

3-(2-Bromophenyl)-1H-pyrazole, 97%

CAS: 114382-20-6 | C9H7BrN2 | 223.073 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H34406.03
also known as H34406-03
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.60
Save 5.95
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.60
Save 5.95
Each
Chemical Identifiers
CAS114382-20-6
IUPAC Name5-(2-bromophenyl)-1H-pyrazole
Molecular FormulaC9H7BrN2
InChI KeyVDHTYVJAQNZWTD-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1C1=CC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay (GC)≥96.0%
Melting Point (clear melt)125.5-134.5?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only